This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt07106872_m1
See other RTP5 GE Assays ›
Gene Symbol:
RTP5
Gene Name:
receptor transporter protein 5 (putative)
Gene Aliases:
C3H2orf85, CXXC11
Chromosome Location:
Chr.3: 120859734 - 120863502 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt07106872_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RTP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC149603.1 - 1 - 2 168 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0