Assay Details
Gene Symbol: dpf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059555.2 | NP_491956.1 | 7 - 8 | 1206 | 75 | |
NM_059556.5 | NP_491957.1 | 7 - 8 | 1194 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
