This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02409564_g1
See other SPE-4 GE Assays ›
Gene Symbol:
spe-4
Gene Name:
Presenilin spe-4
Gene Aliases:
CELE_ZK524.1
Chromosome Location:
Chr.I: 7454660 - 7456953 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02409564_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: spe-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059694.6 NP_492095.1 4 - 5 727 115
GenBank mRNA Z14066.1 - 688 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

membrane protein ectodomain proteolysis
calcium ion transport
organelle organization
Notch signaling pathway
Notch receptor processing
spermatogenesis
spermatid development
protein localization
protein processing
cell differentiation
amyloid precursor protein catabolic process
negative regulation of apoptotic process
spermatid differentiation
beta-amyloid metabolic process
membrane organization
molecular_function
endopeptidase activity
aspartic-type endopeptidase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline