This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02409950_g1
See other ADR-1 GE Assays ›
Gene Symbol:
adr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_H15N14.1
Chromosome Location:
Chr.I: 7773180 - 7777661 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02409950_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: adr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001373102.1 NP_001359658.1 2 - 3 143 75
NM_059752.6 NP_492153.2 2 - 3 159 75
NM_059753.7 NP_492154.2 2 - 3 161 75
NM_182001.4 NP_871801.1 2 - 3 397 75
NM_182003.5 NP_871803.1 2 - 3 139 75
GenBank mRNA AY150819.1 - 2 - 3 143 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

adenosine to inosine editing
RNA processing
mRNA processing
chemotaxis
determination of adult lifespan
regulation of vulval development
positive regulation of chemotaxis
RNA binding
double-stranded RNA binding
double-stranded RNA adenosine deaminase activity
mRNA binding
adenosine deaminase activity
tRNA-specific adenosine deaminase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline