This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02413189_g1
See other MGL-2 GE Assays ›
Gene Symbol:
mgl-2
Gene Name:
protein-coding
Gene Aliases:
CELE_F45H11.4
Chromosome Location:
Chr.I: 10382745 - 10391395 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02413189_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mgl-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001313467.3 NP_001300396.1 2 262 77
NM_001313468.3 NP_001300397.1 2 - 3 341 77
NM_060319.5 NP_492720.3 2 - 3 334 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
G-protein coupled glutamate receptor signaling pathway
regulation of synaptic transmission, glutamatergic
regulation of postsynaptic membrane potential
regulation of postsynaptic cytosolic calcium ion concentration
G-protein coupled receptor activity
glutamate receptor activity
G protein-coupled receptor activity involved in regulation of postsynaptic membrane potential
neurotransmitter receptor activity involved in regulation of postsynaptic cytosolic calcium ion concentration

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline