This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02413192_g1
See other MGL-2 GE Assays ›
Gene Symbol:
mgl-2 maf-1
Gene Name:
protein-coding
bZIP_Maf domain-containing protein
Gene Aliases:
CELE_F45H11.4
CELE_F45H11.6
Chromosome Location:
Chr.I: 10382745 - 10391395 on Build WBcel235
Chromosome Location:
Chr.I: 10382741 - 10391395 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02413192_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mgl-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001313467.3 NP_001300396.1 5 1066 100
NM_001313468.3 NP_001300397.1 5 - 6 1145 100
NM_060319.5 NP_492720.3 5 - 6 1138 100
Gene Symbol: maf-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381197.2 NP_001367180.1 5 - 6 1143 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001381197.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
G-protein coupled glutamate receptor signaling pathway
regulation of synaptic transmission, glutamatergic
regulation of postsynaptic membrane potential
regulation of postsynaptic cytosolic calcium ion concentration
regulation of transcription, DNA-templated
G-protein coupled receptor activity
glutamate receptor activity
G protein-coupled receptor activity involved in regulation of postsynaptic membrane potential
neurotransmitter receptor activity involved in regulation of postsynaptic cytosolic calcium ion concentration
RNA polymerase II core promoter proximal region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline