Assay Details
| Gene Symbol: cdk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026366.3 | NP_001021537.2 | 3 - 4 | 499 | 80 | |
| NM_001350362.3 | NP_001337286.1 | 3 - 4 | 516 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001350362.3 transcripts.
Set Membership: |
|

cdk-2
Genomic Map