Assay Details
| Gene Symbol: unc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026325.2 | NP_001021496.1 | 25 - 26 | 6696 | 86 | |
| NM_001026327.4 | NP_001021498.1 | 9 - 10 | 2230 | 86 | ||
| NM_001262583.3 | NP_001249512.1 | 7 - 8 | 1889 | 86 | ||
| NM_001375256.3 | NP_001361940.1 | 9 - 10 | 2024 | 86 | ||
| GenBank mRNA | AF048834.1 | - | 25 - 26 | 6696 | 86 | |
More Information
Set Membership: |
|

unc-73
Genomic Map