This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02417437_g1
See other PDE-5 GE Assays ›
Gene Symbol:
pde-5
Gene Name:
Probable 3',5'-cyclic phosphodiesterase pde-5
Gene Aliases:
CELE_C32E12.2
Chromosome Location:
Chr.I: 5196073 - 5200263 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02417437_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pde-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059143.4 NP_491544.3 10 - 11 1328 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
signal transduction
phototransduction
chemosensory behavior
determination of adult lifespan
response to alkaline pH
positive regulation of gene expression
negative regulation of cGMP-mediated signaling
microvillus organization
response to hydrogen peroxide
3',5'-cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-474fd:80/100.66.77.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0