This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02423733_m1
See other HDA-3 GE Assays ›
Gene Symbol:
hda-3
Gene Name:
Histone deacetylase
Gene Aliases:
CELE_R06C1.1
Chromosome Location:
Chr.I: 11912791 - 11917719 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02423733_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hda-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_060625.7 NP_493026.1 1 - 2 169 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chromatin organization
histone deacetylation
DNA protection
positive regulation of transcription from RNA polymerase II promoter
histone H3 deacetylation
histone H4 deacetylation
transcription corepressor activity
histone deacetylase activity
hydrolase activity
NAD-dependent histone deacetylase activity (H3-K14 specific)
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-clzgt:80/100.66.76.159:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline