This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02432624_m1
See other MOG-5 GE Assays ›
Gene Symbol:
mog-5
Gene Name:
Probable pre-mRNA-splicing factor ATP-dependent RNA helicase mog-5
Gene Aliases:
CELE_EEED8.5
Chromosome Location:
Chr.II: 5399538 - 5405810 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02432624_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mog-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_062618.7 NP_495019.1 4 - 5 2548 91
GenBank mRNA AF286899.1 - 4 - 5 2555 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

spliceosomal complex disassembly
mRNA processing
multicellular organism development
germ cell development
RNA splicing
embryonic body morphogenesis
feminization of hermaphroditic germ-line
developmental growth
nucleotide binding
nucleic acid binding
RNA binding
RNA helicase activity
helicase activity
protein binding
ATP binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline