Assay Details
| Gene Symbol: sod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026785.4 | NP_001021956.1 | 1 - 2 | 59 | 64 | |
| GenBank mRNA | L20135.1 | - | 1 - 2 | 113 | 64 | |
More Information
Set Membership: |
|

sod-1
Genomic Map