Assay Details
| Gene Symbol: sod-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026785.4 | NP_001021956.1 | 3 - 4 | 305 | 81 | |
| NM_001026786.5 | NP_001021957.1 | 2 - 3 | 232 | 81 | ||
| GenBank mRNA | L20135.1 | - | 3 - 4 | 359 | 81 | |
More Information
Set Membership: |
|

sod-1
Genomic Map