This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02444754_g1
See other GSR-1 GE Assays ›
Gene Symbol:
gsr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C46F11.2
Chromosome Location:
Chr.III: 3650905 - 3653404 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02444754_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gsr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026049.6 NP_001021220.1 4 - 5 1195 67
NM_001384057.1 4 - 5 1151 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to superoxide
glutathione metabolic process
electron transport chain
cellular response to oxidative stress
ecdysis, collagen and cuticulin-based cuticle
cell redox homeostasis
oxidation-reduction process
cellular oxidant detoxification
glutathione-disulfide reductase activity
electron carrier activity
oxidoreductase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline