Assay Details
| Gene Symbol: adr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066193.5 | NP_498594.1 | 1 - 2 | 405 | 133 | |
| GenBank mRNA | AF051275.1 | - | 1 - 2 | 427 | 133 | |
| Gene Symbol: R151.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382910.1 | 1 - 2 | 407 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382910.1 transcripts.
Set Membership: |
|

adr-2
Genomic Map