This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02449980_g1
See other MET-2 GE Assays ›
Gene Symbol:
met-2
Gene Name:
Histone-lysine N-methyltransferase met-2
Gene Aliases:
CELE_R05D3.11
Chromosome Location:
Chr.III: 8375300 - 8380863 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02449980_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: met-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066447.6 NP_498848.4 1 - 2 94 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
sex differentiation
histone H3-K36 methylation
negative regulation of gene expression
cell differentiation
methylation
histone lysine methylation
negative regulation of vulval development
regulation of gene expression, epigenetic
negative regulation of meiotic nuclear division
meiotic cell cycle
histone H3-K9 methylation
heterochromatin organization
vulval cell fate commitment
positive regulation of methylation-dependent chromatin silencing
DNA binding
methyltransferase activity
zinc ion binding
transferase activity
histone-lysine N-methyltransferase activity
metal ion binding
histone methyltransferase activity (H3-K9 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-wtg8n:80/100.66.76.99:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline