This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02452471_g1
See other RIB-2 GE Assays ›
Gene Symbol:
rib-2
Gene Name:
Exostosin-2 homolog
Gene Aliases:
CELE_K01G5.6
Chromosome Location:
Chr.III: 10742777 - 10746675 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02452471_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rib-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_066967.8 NP_499368.1 7 - 8 2198 116
GenBank mRNA AB077851.1 - 7 - 8 2118 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
protein glycosylation
heparan sulfate proteoglycan biosynthetic process
embryonic morphogenesis
pharynx development
glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
protein binding
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
enzyme binding
heparan sulfate N-acetylglucosaminyltransferase activity
metal ion binding
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0