This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02456685_g1
See other SYX-4 GE Assays ›
Gene Symbol:
syx-4
Gene Name:
Putative syntaxin-4
Gene Aliases:
CELE_T01B11.3
Chromosome Location:
Chr.IV: 8457280 - 8458933 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02456685_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: syx-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_069038.4 NP_501439.2 3 - 4 359 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter transport
intracellular protein transport
exocytosis
vesicle fusion
synaptic vesicle docking
vesicle-mediated transport
regulation of exocytosis
synaptic vesicle fusion to presynaptic active zone membrane
positive regulation of cytokinesis
vesicle docking
cortical granule exocytosis
SNARE binding
SNAP receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0