This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02478534_g1
See other NATC-1 GE Assays ›
Gene Symbol:
natc-1
Gene Name:
N-alpha-AcetylTransferase C complex subunit
Gene Aliases:
CELE_T23B12.4
Chromosome Location:
Chr.V: 8462327 - 8465569 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02478534_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: natc-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_072778.7 NP_505179.1 6 - 7 2208 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
determination of adult lifespan
response to heat
N-terminal peptidyl-methionine acetylation
dauer larval development
response to paraquat
stress response to copper ion
stress response to cadmium ion
stress response to zinc ion
peptide alpha-N-acetyltransferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-695c48667f-m64s4:80/100.66.74.235:80.
git-commit: c086736f0d622c41bbf3ea4d71b1c2b8619a5b9d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.35.0-Offline