This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02488017_g1
See other SRZ-96 GE Assays ›
Gene Symbol:
srz-96
Gene Name:
Serpentine Receptor, class Z
Gene Aliases:
CELE_Y68A4A.6
Chromosome Location:
Chr.V: 17199226 - 17201757 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02488017_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: srz-96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_074955.2 NP_507356.1 1 - 2 339 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-774fdbf798-5f8tx:80/100.66.79.126:80.
git-commit: 7dd85a2aaed5e3122cf027567276b2918d4b9fce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.0-Offline