This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02488621_m1
See other DPY-21 GE Assays ›
Gene Symbol:
dpy-21
Gene Name:
Lysine-specific demethylase 9
Gene Aliases:
CELE_Y59A8B.1
Chromosome Location:
Chr.V: 17936282 - 17949193 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02488621_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dpy-21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029095.4 NP_001024266.1 6 - 7 3961 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity
metal ion binding
dioxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-mt4f6:80/100.66.78.48:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0