This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf00976241_m1
See other ESRRG GE Assays ›
Gene Symbol:
ESRRG
Gene Name:
estrogen related receptor gamma
Gene Aliases:
-
Chromosome Location:
Chr.38: 11825055 - 12439143 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf00976241_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESRRG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022415393.1 XP_022271101.1 3 - 4 766 62
XM_022415394.1 XP_022271102.1 3 - 4 648 62
XM_022415395.1 XP_022271103.1 6 - 7 1086 62
XM_022415396.1 XP_022271104.1 7 - 8 1281 62
XM_022415397.1 XP_022271105.1 5 - 6 919 62
XM_022415398.1 XP_022271106.1 6 - 7 954 62
XM_022415399.1 XP_022271107.1 5 - 6 863 62
XM_022415400.1 XP_022271108.1 5 - 6 824 62
XM_022415401.1 XP_022271109.1 6 - 7 1030 62
XM_022415402.1 XP_022271110.1 6 - 7 1140 62
XM_022415403.1 XP_022271111.1 6 - 7 1031 62
XM_022415404.1 XP_022271112.1 5 - 6 838 62
XM_022415405.1 XP_022271113.1 5 - 6 945 62
XM_022415406.1 XP_022271114.1 5 - 6 1003 62
XM_022415407.1 XP_022271115.1 5 - 6 871 62
XM_022415408.1 XP_022271116.1 3 - 4 652 62
XM_844183.5 XP_849276.1 3 - 4 764 62

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0