This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf01061978_m1
See other CNTFR GE Assays ›
Gene Symbol:
CNTFR
Gene Name:
ciliary neurotrophic factor receptor
Gene Aliases:
CNTFRALPHA
Chromosome Location:
Chr.11: 51226951 - 51266431 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf01061978_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNTFR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003353.1 NP_001003353.1 3 - 4 343 106
XM_005626374.3 XP_005626431.1 4 - 5 558 106
XM_005626376.3 XP_005626433.1 4 - 5 518 106
XM_005626377.3 XP_005626434.1 4 - 5 562 106
GenBank mRNA AF529215.1 - 3 - 4 343 106

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

suckling behavior
brainstem development
sex differentiation
positive regulation of cell proliferation
negative regulation of neuron apoptotic process
skeletal muscle organ development
receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline