This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02622093_m1
See other UCP1 GE Assays ›
Gene Symbol:
UCP1
Gene Name:
uncoupling protein 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 2254881 - 2289902 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02622093_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003046.1 NP_001003046.1 818 73
GenBank mRNA AB046106.1 - 818 73
AF201376.1 - 389 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

response to temperature stimulus
response to nutrient levels
cellular response to hormone stimulus
cellular response to reactive oxygen species
cellular response to fatty acid
hydrogen ion transmembrane transport
regulation of reactive oxygen species biosynthetic process
mitochondrial transmembrane transport
adaptive thermogenesis
oxidative phosphorylation uncoupler activity
purine ribonucleotide binding
long-chain fatty acid binding
cardiolipin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0