This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02623929_m1
See other IL1RN GE Assays ›
Gene Symbol:
IL1RN
Gene Name:
interleukin 1 receptor antagonist
Gene Aliases:
IL-1ra
Chromosome Location:
Chr.17: 37236690 - 37250424 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02623929_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL1RN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003096.2 NP_001003096.1 2 - 3 258 84
XM_022404316.1 XP_022260024.1 3 - 4 283 84
XM_022404317.1 XP_022260025.1 3 - 4 496 84
GenBank mRNA AF216526.1 - 2 - 3 224 84
AY026462.2 - 2 - 3 252 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris Inventoried Probe spans exons

Panther Classification:

Molecular Function -

interleukin superfamily

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
signal transduction
insulin secretion
negative regulation of heterotypic cell-cell adhesion
response to glucocorticoid
cytokine activity
interleukin-1, Type I receptor binding
interleukin-1, Type II receptor binding
interleukin-1 Type I receptor antagonist activity
interleukin-1 Type II receptor antagonist activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0