This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02624992_m1
See other EPB41L5 GE Assays ›
Gene Symbol:
EPB41L5
Gene Name:
erythrocyte membrane protein band 4.1 like 5
Gene Aliases:
BE37
Chromosome Location:
Chr.19: 29916994 - 30063243 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02624992_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41L5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002952.1 NP_001002952.1 7 - 8 636 62
XM_005631761.2 XP_005631818.1 7 - 8 662 62
XM_014121360.2 XP_013976835.1 7 - 8 662 62
XM_014121361.2 XP_013976836.1 7 - 8 721 62
XM_014121362.2 XP_013976837.1 7 - 8 663 62
XM_014121364.2 XP_013976839.1 7 - 8 663 62
XM_014121366.2 XP_013976841.1 7 - 8 662 62
XM_022405860.1 XP_022261568.1 7 - 8 662 62
GenBank mRNA AJ401272.1 - 7 - 8 636 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Gene Ontology Categories:

Process(es)

cytoskeletal protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9mkk8:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0