This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02625201_s1
See other GJA5 GE Assays ›
Gene Symbol:
GJA5
Gene Name:
gap junction protein alpha 5
Gene Aliases:
Cx40
Chromosome Location:
Chr.17: 58289327 - 58305315 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02625201_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJA5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017442.2 NP_001017442.2 2 654 78
GenBank mRNA AY567709.1 - 1 562 78
DQ666281.1 - 1 558 78
EF589114.1 - 2 654 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
angiogenesis
renal system process involved in regulation of systemic arterial blood pressure
outflow tract morphogenesis
cardiac conduction system development
mitral valve development
pulmonary valve formation
septum primum development
atrial ventricular junction remodeling
gap junction assembly
embryonic limb morphogenesis
embryonic heart tube development
foramen ovale closure
artery morphogenesis
transmembrane transport
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of atrial cardiac muscle cell membrane depolarization
regulation of ventricular cardiac muscle cell membrane depolarization
ventricular septum morphogenesis
atrial septum morphogenesis
ventricular cardiac muscle cell action potential
SA node cell action potential
atrial cardiac muscle cell to AV node cell communication by electrical coupling
AV node cell to bundle of His cell communication by electrical coupling
bundle of His cell to Purkinje myocyte communication by electrical coupling
regulation of heart rate by cardiac conduction
regulation of blood vessel diameter
regulation of AV node cell action potential
regulation of bundle of His cell action potential
regulation of Purkinje myocyte action potential
regulation of atrial cardiac muscle cell action potential
regulation of renin secretion into blood stream
regulation of membrane depolarization during cardiac muscle cell action potential
gap junction hemi-channel activity
gap junction channel activity involved in atrial cardiac muscle cell-AV node cell electrical coupling
gap junction channel activity involved in AV node cell-bundle of His cell electrical coupling
gap junction channel activity involved in bundle of His cell-Purkinje myocyte electrical coupling

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline