This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02625569_m1
See other COMMD1 GE Assays ›
Gene Symbol:
COMMD1
Gene Name:
copper metabolism domain containing 1
Gene Aliases:
MURR1
Chromosome Location:
Chr.10: 61886281 - 62059610 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02625569_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COMMD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003055.1 NP_001003055.1 1 - 2 200 71
GenBank mRNA AX786408.1 - 1 - 2 200 71
AY047597.1 - 1 - 2 200 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein transport
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
regulation of proteasomal ubiquitin-dependent protein catabolic process
copper ion homeostasis
copper ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0