This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02626372_g1
See other AGER GE Assays ›
Gene Symbol:
AGER
Gene Name:
advanced glycosylation end-product specific receptor
Gene Aliases:
RAGE
Chromosome Location:
Chr.12: 1574251 - 1577410 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02626372_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGER
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001048081.2 NP_001041546.2 3 - 4 377 63
GenBank mRNA AY382178.1 - 2 - 3 281 63
AY836152.1 - 3 - 4 377 63
DQ125936.1 - 3 - 4 359 63
DQ125937.1 - 3 - 4 359 63
DQ125938.1 - 3 - 4 359 63
DQ125939.1 - 3 - 4 359 63
DQ125940.1 - 3 - 4 359 63
DQ125941.1 - 3 - 4 359 63
EU428787.1 - 2 - 3 529 63
EU428788.1 - 1 - 2 660 63
EU428789.1 - 3 - 4 359 63
EU428790.1 - 1 - 2 660 63
EU428791.1 - 2 - 3 490 63
EU428792.1 - 1 - 2 660 63
EU428793.1 - 2 - 3 529 63
EU428794.1 - 2 - 3 529 63
EU428796.1 - 3 - 4 359 63
EU428797.1 - 2 - 3 529 63
EU428798.1 - 2 - 3 529 63
EU428799.1 - 2 - 3 529 63
EU428800.1 - 2 - 3 529 63
EU428801.1 - 2 - 3 529 63
EU428802.1 - 1 - 2 660 63
EU428803.1 - 2 - 3 529 63
EU428805.1 - 1 - 2 660 63
EU428806.1 - 2 - 3 529 63
EU428807.1 - 2 - 3 529 63
EU428808.1 - 2 - 3 529 63
EU428809.1 - 2 - 3 529 63
EU428810.1 - 1 - 2 660 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0