This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02626404_m1
See other UNK GE Assays ›
Gene Symbol:
UNK
Gene Name:
unk zinc finger
Gene Aliases:
ZC3H5
Chromosome Location:
Chr.9: 4767696 - 4801324 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02626404_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003390.1 NP_001003390.1 898 75
XM_022422543.1 XP_022278251.1 2104 75
XM_022422544.1 XP_022278252.1 1425 75
XM_022422545.1 XP_022278253.1 1664 75
XM_022422546.1 XP_022278254.1 3602 75
XM_022422547.1 XP_022278255.1 964 75
XM_022422548.1 XP_022278256.1 2000 75
XM_022422549.1 XP_022278257.1 964 75
XM_022422551.1 XP_022278259.1 961 75
XM_022422552.1 XP_022278260.1 963 75
XM_022422553.1 XP_022278261.1 987 75
XM_022422554.1 XP_022278262.1 2196 75
XM_022422555.1 XP_022278263.1 2171 75
XM_022422556.1 XP_022278264.1 2165 75
XM_022422557.1 XP_022278265.1 1834 75
XM_022422558.1 XP_022278266.1 1928 75
XM_022422559.1 XP_022278267.1 1579 75
GenBank mRNA AY422569.1 - 898 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
cell morphogenesis involved in neuron differentiation
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline