This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02626832_m1
See other HSD3B2 GE Assays ›
Gene Symbol:
HSD3B2
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2
Gene Aliases:
3b-HSD, HSD3B, HSD3B1
Chromosome Location:
Chr.17: 56552945 - 56561062 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02626832_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD3B2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001010954.2 NP_001010954.1 1 - 2 49 102
GenBank mRNA AY739720.1 - 1 - 2 50 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001010954.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
oxidation-reduction process
3-beta-hydroxy-delta5-steroid dehydrogenase activity
steroid delta-isomerase activity
cholesterol dehydrogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0