This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02627675_m1
See other ADGRE3 GE Assays ›
Gene Symbol:
ADGRE3
Gene Name:
adhesion G protein-coupled receptor E3
Gene Aliases:
EMR3
Chromosome Location:
Chr.20: 48021336 - 48060618 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02627675_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRE3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038666.1 NP_001033755.1 8 - 9 987 69
XM_022406246.1 XP_022261954.1 8 - 9 1020 69
GenBank mRNA DQ227277.1 - 8 - 9 987 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-nn99n:80/100.66.79.68:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0