This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02633444_m1
See other TTPAL GE Assays ›
Gene Symbol:
TTPAL
Gene Name:
alpha tocopherol transfer protein like
Gene Aliases:
-
Chromosome Location:
Chr.24: 31920499 - 31958799 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02633444_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTPAL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005635090.3 XP_005635147.1 3 - 4 626 64
XM_005635091.3 XP_005635148.1 2 - 3 567 64
XM_014106891.2 XP_013962366.1 2 - 3 753 64
XM_014106892.2 XP_013962367.1 3 - 4 784 64
XM_022409463.1 XP_022265171.1 2 - 3 597 64
XM_534426.6 XP_534426.1 2 - 3 547 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0