This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02634102_mH
See other NAPB GE Assays ›
Gene Symbol:
NAPB
Gene Name:
NSF attachment protein beta
Gene Aliases:
-
Chromosome Location:
Chr.24: 217287 - 258278 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02634102_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAPB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014107169.2 XP_013962644.1 2 - 3 272 72
XM_014107170.2 XP_013962645.1 2 - 3 272 72
XM_014107171.2 XP_013962646.2 2 - 3 563 72
XM_022408880.1 XP_022264588.1 2 - 3 256 72
XM_022408881.1 XP_022264589.1 3 - 4 324 72
XM_022408882.1 XP_022264590.1 2 - 3 560 72
XM_022408884.1 XP_022264592.1 3 - 4 324 72
XM_534322.6 XP_534322.2 2 - 3 270 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_014107171.2, XM_022408884.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0