This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02634447_m1
See other LIPJ GE Assays ›
Gene Symbol:
LIPJ
Gene Name:
lipase family member J
Gene Aliases:
-
Chromosome Location:
Chr.26: 38419821 - 38441234 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02634447_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPJ
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005636643.3 XP_005636700.1 5 - 6 622 72
XM_005636644.3 XP_005636701.1 5 - 6 1006 72
XM_022410312.1 XP_022266020.1 5 - 6 820 72
XM_022410313.1 XP_022266021.1 5 - 6 820 72
XM_022410314.1 XP_022266022.1 5 - 6 820 72
XM_022410315.1 XP_022266023.1 5 - 6 820 72
XM_534779.6 XP_534779.4 5 - 6 820 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0