Assay Details
| Gene Symbol: CD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616714.3 | XP_005616771.3 | 4 - 5 | 780 | 80 | |
| XM_014115358.2 | XP_013970833.1 | 4 - 5 | 550 | 80 | ||
| XM_014115369.2 | XP_013970844.1 | 3 - 4 | 718 | 80 | ||
| XM_022403552.1 | XP_022259260.1 | 4 - 5 | 806 | 80 | ||
| XM_862798.4 | XP_867891.3 | 4 - 5 | 783 | 80 | ||
More Information
Set Membership: |
|

CD22
Genomic Map