This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02641341_m1
See other TAP2 GE Assays ›
Gene Symbol:
TAP2
Gene Name:
transporter 2, ATP binding cassette subfamily B member
Gene Aliases:
APT2
Chromosome Location:
Chr.12: 2407512 - 2417469 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02641341_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284495.1 NP_001271424.1 5 - 6 984 73
XM_005627192.3 XP_005627249.1 5 - 6 1122 73
XM_005627193.3 XP_005627250.1 5 - 6 1091 73
GenBank mRNA JN656400.2 - 5 - 6 987 73
JN656401.1 - 4 - 5 840 73
JN656402.1 - 4 - 5 840 73
JN656403.1 - 4 - 5 840 73
JN656404.1 - 4 - 5 840 73
JN656405.1 - 4 - 5 840 73
JN656406.1 - 4 - 5 840 73
JN656407.1 - 4 - 5 840 73
JN656408.1 - 4 - 5 840 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0