This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02643059_m1
See other GPR1 GE Assays ›
Gene Symbol:
GPR1
Gene Name:
G protein-coupled receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.37: 14725573 - 14776009 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02643059_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005640575.3 XP_005640632.1 3 - 4 418 82
XM_005640576.3 XP_005640633.1 3 - 4 417 82
XM_005640578.3 XP_005640635.1 3 - 4 223 82
XM_005640581.3 XP_005640638.1 4 - 5 1654 82
XM_005640582.3 XP_005640639.1 2 - 3 340 82
XM_005640583.3 XP_005640640.1 2 - 3 342 82
XM_014111130.2 XP_013966605.1 4 - 5 551 82
XM_022415109.1 XP_022270817.1 4 - 5 298 82
XM_022415110.1 XP_022270818.1 4 - 5 543 82
XM_022415111.1 XP_022270819.1 3 - 4 236 82
XM_022415112.1 XP_022270820.1 3 - 4 286 82
XM_022415113.1 XP_022270821.1 3 - 4 222 82
XM_022415114.1 XP_022270822.1 3 - 4 583 82
XM_022415116.1 XP_022270824.1 4 - 5 1628 82
XM_022415117.1 XP_022270825.1 2 - 3 1452 82
XM_022415118.1 XP_022270826.1 2 - 3 210 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005640575.3, XM_005640576.3, XM_005640578.3, XM_005640581.3, XM_005640582.3, XM_005640583.3, XM_014111130.2, XM_022415109.1, XM_022415110.1, XM_022415111.1, XM_022415112.1, XM_022415113.1, XM_022415114.1, XM_022415116.1, XM_022415117.1, XM_022415118.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline