This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02648802_m1
See other TEAD2 GE Assays ›
Gene Symbol:
TEAD2
Gene Name:
TEA domain transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 107115014 - 107130563 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02648802_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TEAD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005616318.3 XP_005616375.1 8 - 9 711 82
XM_005616319.3 XP_005616376.1 8 - 9 701 82
XM_014118921.1 XP_013974396.1 6 - 7 593 82
XM_014118922.1 XP_013974397.1 7 - 8 605 82
XM_014118924.1 XP_013974399.1 7 - 8 605 82
XM_022423898.1 XP_022279606.1 8 - 9 722 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0