This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02649068_m1
See other LRTOMT GE Assays ›
Gene Symbol:
LRTOMT
Gene Name:
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.21: 25987687 - 26000545 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02649068_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRTOMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282031.1 NP_001268960.1 4 - 5 402 59
XM_014122427.2 XP_013977902.1 4 - 5 492 59
XM_014122428.2 XP_013977903.1 4 - 5 485 59
XM_014122429.2 XP_013977904.1 3 - 4 330 59
XM_014122430.2 XP_013977905.1 3 - 4 375 59
XM_014122432.2 XP_013977907.1 2 - 3 234 59
XM_022407435.1 XP_022263143.1 2 - 3 264 59
XM_022407436.1 XP_022263144.1 2 - 3 246 59
GenBank mRNA CX985118.1 - 3 - 4 574 59
DN342337.1 - 4 - 5 402 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline