This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02649070_m1
See other LRTOMT GE Assays ›
Gene Symbol:
LRTOMT
Gene Name:
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.21: 25987687 - 26000545 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02649070_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRTOMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282031.1 NP_001268960.1 6 - 7 756 80
XM_014122427.2 XP_013977902.1 6 - 7 846 80
XM_014122428.2 XP_013977903.1 6 - 7 839 80
XM_014122429.2 XP_013977904.1 5 - 6 684 80
XM_014122430.2 XP_013977905.1 5 - 6 729 80
XM_014122432.2 XP_013977907.1 4 - 5 588 80
XM_022407435.1 XP_022263143.1 4 - 5 618 80
XM_022407436.1 XP_022263144.1 4 - 5 600 80
XM_022407437.1 XP_022263145.1 3 - 4 377 80
GenBank mRNA CO603444.1 - 1 - 2 227 80
CX985118.1 - 1 - 2 220 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline