This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02651081_m1
See other CHST15 GE Assays ›
Gene Symbol:
CHST15
Gene Name:
carbohydrate sulfotransferase 15
Gene Aliases:
-
Chromosome Location:
Chr.28: 33427861 - 33490350 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02651081_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHST15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005637824.3 XP_005637881.1 5 - 6 1772 66
XM_005637826.3 XP_005637883.1 5 - 6 1966 66
XM_005637827.3 XP_005637884.1 5 - 6 1815 66
XM_022411597.1 XP_022267305.1 6 - 7 2093 66
XM_022411598.1 XP_022267306.1 4 - 5 1668 66
XM_022411599.1 XP_022267307.1 5 - 6 1989 66
XM_544058.7 XP_544058.4 4 - 5 1862 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline