This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02651167_m1
See other SDSL GE Assays ›
Gene Symbol:
SDSL
Gene Name:
serine dehydratase like
Gene Aliases:
-
Chromosome Location:
Chr.26: 10731972 - 10759768 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02651167_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDSL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005636263.3 XP_005636320.1 864 81
XM_005636266.3 XP_005636323.1 519 81
XM_005636268.3 XP_005636325.1 863 81
XM_014108039.2 XP_013963514.1 661 81
XM_014108040.2 XP_013963515.1 585 81
XM_022410256.1 XP_022265964.1 1067 81
XM_022410257.1 XP_022265965.1 870 81
XM_022410258.1 XP_022265966.1 786 81
XM_022410259.1 XP_022265967.1 860 81
XM_022410260.1 XP_022265968.1 858 81
XM_022410261.1 XP_022265969.1 657 81
XM_022410262.1 XP_022265970.1 788 81
XM_022410263.1 XP_022265971.1 682 81
XM_022410264.1 XP_022265972.1 539 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rw9fd:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0