This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02652650_m1
See other UVSSA GE Assays ›
Gene Symbol:
UVSSA
Gene Name:
UV stimulated scaffold protein A
Gene Aliases:
-
Chromosome Location:
Chr.3: 62647992 - 62674830 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02652650_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UVSSA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014112584.2 XP_013968059.1 5 - 6 1172 68
XM_022417136.1 XP_022272844.1 5 - 6 1172 68
XM_022417137.1 XP_022272845.1 5 - 6 1243 68
XM_022417139.1 XP_022272847.1 5 - 6 1383 68
XM_022417140.1 XP_022272848.1 5 - 6 1159 68
XM_022417141.1 XP_022272849.1 5 - 6 1157 68
XM_022417142.1 XP_022272850.1 4 - 5 891 68
XM_022417143.1 XP_022272851.1 5 - 6 1223 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0