This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02655188_m1
See other SATL1 GE Assays ›
Gene Symbol:
SATL1
Gene Name:
spermidine/spermine N1-acetyl transferase like 1
Gene Aliases:
-
Chromosome Location:
Chr.X: 65955694 - 65984081 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02655188_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SATL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005641536.3 XP_005641593.1 2 - 3 1083 115
XM_005641537.3 XP_005641594.1 2 - 3 1083 115
XM_022416116.1 XP_022271824.1 2 - 3 1085 115
XM_549112.5 XP_549112.3 2 - 3 1083 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0