Assay Details
| Gene Symbol: SATL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641536.3 | XP_005641593.1 | 4 - 5 | 1203 | 122 | |
| XM_022416116.1 | XP_022271824.1 | 5 - 6 | 1259 | 122 | ||
| XM_549112.5 | XP_549112.3 | 5 - 6 | 1305 | 122 | ||
More Information
Set Membership: |
|

SATL1
Genomic Map