This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02655327_m1
See other SPNS3 GE Assays ›
Gene Symbol:
SPNS3
Gene Name:
sphingolipid transporter 3 (putative)
Gene Aliases:
-
Chromosome Location:
Chr.5: 30464288 - 30493609 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02655327_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPNS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014113477.2 XP_013968952.1 10 - 11 1613 103
XM_014113478.2 XP_013968953.1 10 - 11 1598 103
XM_014113479.2 XP_013968954.1 11 - 12 1494 103
XM_014113480.2 XP_013968955.1 11 - 12 1490 103
XM_022418398.1 XP_022274106.1 11 - 12 1427 103
XM_022418399.1 XP_022274107.1 8 - 9 1394 103
XM_843290.5 XP_848383.2 10 - 11 1403 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-m2kmk:80/100.66.77.245:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0