This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02655510_m1
See other TVP23B GE Assays ›
Gene Symbol:
TVP23B
Gene Name:
trans-golgi network vesicle protein 23 homolog B
Gene Aliases:
FAM18B1
Chromosome Location:
Chr.5: 39079975 - 39099309 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02655510_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TVP23B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001281930.1 NP_001268859.1 5 - 6 533 60
XM_005619526.3 XP_005619583.1 5 - 6 633 60
XM_022418033.1 XP_022273741.1 5 - 6 547 60
XM_022418034.1 XP_022273742.1 5 - 6 564 60
XM_022418035.1 XP_022273743.1 5 - 6 468 60
GenBank mRNA DN869649.1 - 5 - 6 504 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0