Assay Details
| Gene Symbol: PIGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109446.2 | XP_013964921.1 | 11 - 12 | 1512 | 85 | |
| XM_022412646.1 | XP_022268354.1 | 11 - 12 | 1515 | 85 | ||
More Information
Set Membership: |
|

PIGB
Genomic Map