This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02659462_m1
See other RIC3 GE Assays ›
Gene Symbol:
RIC3
Gene Name:
RIC3 acetylcholine receptor chaperone
Gene Aliases:
-
Chromosome Location:
Chr.21: 31629824 - 31681832 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02659462_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005633628.3 XP_005633685.1 3 - 4 477 86
XM_005633629.3 XP_005633686.1 3 - 4 476 86
XM_022407798.1 XP_022263506.1 2 - 3 950 86
XM_022407799.1 XP_022263507.1 2 - 3 867 86
XM_022407800.1 XP_022263508.1 2 - 3 368 86
XM_022407803.1 XP_022263511.1 3 - 4 496 86
XM_022407804.1 XP_022263512.1 3 - 4 489 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0